Slot samplers
Supplier: Sampling Systems
Certificates
About this item
These all layer samplers have a robust all stainless steel construction and are suitable for sampling free flowing powders, granules and even slightly cohesive powders.
- Multilevel samplers
- Cross section type
- With or without bottle holder
For taking a large volume, cross sectional sample from a container.
Ordering information: Sample collecting bottles (for use with the sampler with bottle holder) as well as cleaning brushes can be ordered separately.
Frequently Bought Together