Order Entry
Switzerland
ContactUsLinkComponent
Glycoprotein stain
Glycoprotein stain
Catalog # 786-254
Supplier:  G-Biosciences
CAS Number:  
undefined
Glycoprotein stain
Catalog # 786-254
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Glycoprotein stain for10 mini gels (8×8 cm) or 20 nitrocellulose membrane (8×8 cm)

Specifications

About this item

The stain uses an enhanced Periodic Acid-Schiff (PAS) method for detection of glycoprotein sugars. The supplied agent first oxidises the cis-diol sugar groups to aldehydes. The aldehyde groups react with the sensitive glycoprotein stain solution forming Schiff bonds and producing strong magenta colour bands. RAPIDstain™ can be used after glycoprotein staining to detect non-glycosylated proteins and the use of the stain enhances glycoprotein staining. The glycoprotein stainis highly convenient as all the key reagents required for staining are supplied and a unique positive & negative control is included.

  • Specific glycosylated protein staining with optional non-glycosylated protein staining
  • Two stains in one kit
  • Nanogram level sensitivity
  • Stable periodate/Schiff staining

Suitable for the highly sensitive detection of glycoproteins following gel electrophoresis or protein transfer to nitrocellulose membranes.

Delivery: Supplied with RAPIDstain™, an enhanced Coomassie stain.
Caution: Store Glyco‐positive and negative control at -20 °C and other components at 4 °C (Shipped at ambient temperature)