Order Entry
Switzerland
ContactUsLinkComponent
 
Reversed phase chromatography columns, Asahipak preparative type
  SHOWF6820001
 :  Resonac
Reversed phase chromatography columns, Asahipak preparative type
  SHOWF6820001
 :  Resonac

 

  • Column Format:
    Preparative
  • Flow rate:
    3,0 ml/min
  • Ø int.:
    10,00 mm
  • Particle Size:
    5 µm
  • Pore size:
    250 Å
  • Max. pressure:
    15 MPa (150 bar)
  • Description:
    Asahipak ODP-50 10E
  • Length:
    250 mm
  • Seq:
    02
  • USP code:
    L67
  • pH range:
    2 - 13
  • Temperature limit:
    4 - 60 °C
  • Carbon load:
    17%
  • Column hardware:
    Stainless steel
  • Pk:
    1

 

 

Polymer-based columns usable in 100% water and buffer solutions. Ideal for the analysis of basic substances.

  • Wide pH range (2 to 13)
  • Suitable for the analysis of proteins, peptides and amino acids