Platinum electrodes, Tweezertrodes™
Supplier: BTX
Certificates
About this item
Tweezertrodes™ are reusable, tweezer style electrodes for in vivo, and in utero drug or gene delivery. Tweezertrodes™ consist of a standard 12 cm tweezer that has been modified with stainless steel or platinum circular electrodes at the tip. The gap between the electrode disks may be adjusted from under 1 mm to over 2 cm.
- May be cleaned with a mild detergent and sterilised with ethanol or ethylene oxide
- Can be connected to an electroporator via a connection cable
- Compatible with the BTX ECM® 630, 830 and 2001
- Ideal for in vivo and in utero applications in animal tissues
Frequently Bought Together