ContactUsLinkComponent
Multimode columns, Asahipak GS-HQ series preparative type
Multimode columns, Asahipak GS-HQ series preparative type
Supplier:  Resonac
Certificates
Multimode columns, Asahipak GS-HQ series preparative type
Supplier:  Resonac
Specifications
  • Catalog No:
  • SHOWF6810035
  • SHOWF6810034
  • Description:
  • Asahipak GS-320 20G
  • Asahipak GS-220 20G
  • pH range:
  • 2 - 12
  • 2 - 9
  • Temperature limit:
  • 4 - 60 °C
  • 4 - 60 °C