Latex gloves, Vasco® Sensitive
Supplier: B. Braun
Certificates
About this item
These ambidextrous gloves are ideal for medical examinations and for non hazardous applications in laboratories.
- Non sterile, powder-free
- Polymer coating inside for easier donning
- Straight fingers, ambidextrous fitting
- Rolled rim
- Micro-rough surface, textured fingers
Certifications: EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS; Cat. III; AQL 1,5
Food contact approved
Food contact approved
Specifications
- Catalog No:
- BRAU6067565
- BRAU6067526
- BRAU6067590
- BRAU6067549
- BRAU6067500
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Cuff thickness:
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- Finger thickness:
- 0,125 mm
- 0,125 mm
- 0,125 mm
- 0,125 mm
- 0,125 mm
- Palm thickness:
- 0,11 mm
- 0,11 mm
- 0,11 mm
- 0,11 mm
- 0,11 mm
- EN norm:
- EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
- EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
- EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
- EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
- EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
- Food contact approved:
- +
- +
- +
- +
- +
Specifications
Frequently Bought Together