Orders LinkContactUsLinkComponent
Drums, combination
Drums, combination
Supplier:   DUPERTHAL
Drums, combination
Supplier:  DUPERTHAL
Certificates
Drums, combination
Drums, combination
2 Options Available Below
Product Details & Documents
These combination drum has a outer jacket steel sheet with blue enamel and inner bubble polyethylene.
  • Tight closure
  • BAM allowed
  • Transport approval for all packaging groups
Container identification according to GefStoffV and BGV A8.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 6 L
    250 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53.60 SFr.
    53.60 SFr.
    1 * 1 ST
       53.60 SFr.
  • 12 L
    250 mm
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58.70 SFr.
    58.70 SFr.
    1 * 1 ST
       58.70 SFr.