Anion exchange chromatography columns, IEC series
: Resonac
QA-825, DEAE-825: Suitable for the analysis of relatively high molecular weight compounds, such as proteins, peptides, DNA, and RNA. Usable across wide pH range between 2 and 12.
DEAE3N-4T: Non porous base material. For rapid analysis.
- Catalog No:
- 554-2857
- 554-2853
- Max. pressure:
- 2,0 MPa (20 bar)
- 2,0 MPa (20 bar)
- pH range:
- 2 - 12
- 2 - 12
- Temperature limit:
- 10 - 50 °C
- 10 - 50 °C
- USP code:
- L23
- L23
Frequently Bought Together