Orders LinkContactUsLinkComponent
Density thermo-hydrometers
Density thermo-hydrometers
Supplier:   Alla France
Density thermo-hydrometers
Supplier:  Alla France
Certificates
Density thermo-hydrometers
Density thermo-hydrometers
6 Options Available Below
Product Details & Documents
General purpose thermo-hydrometers with red liquid filling and reading below meniscus.
Product Family Options
Product Information
  • Range
    Division
    Temperature Range
    Length
    Availability
    Price
  • 0,800 - 0,860 g/ml
    0,001 g/ml
    0…40 °C
    280 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 835.00
    SFr. 835.00
    1 * 5 ST
       SFr. 835.00
  • 0,950 - 1,010 g/ml
    0,001 g/ml
    0…40 °C
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 179.00
    SFr. 179.00
    1 * 1 ST
       SFr. 179.00
  • 1,000 - 1,060 g/ml
    0,001 g/ml
    0…40 °C
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 195.00
    SFr. 195.00
    1 * 1 ST
       SFr. 195.00
  • 1,050 - 1,110 g/ml
    0,001 g/ml
    0…40 °C
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 195.00
    SFr. 195.00
    1 * 1 ST
       SFr. 195.00
  • 1,110 - 1,160 g/ml
    0,001 g/ml
    0…40 °C
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 195.00
    SFr. 195.00
    1 * 1 ST
       SFr. 195.00
  • 1,500 - 1,600 g/ml
    0,001 g/ml
    0…40 °C
    310 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 98.80
    SFr. 98.80
    1 * 1 ST
       SFr. 98.80