Hydrometers A.S.T.M. E100 for mineral oils
Anbieter: Alla France
Zertifikate
Über diesen Artikel
The scale of the hydrometer is fully marked with all the reference norms, the calibration temperature, the surface tension, the individual number of the instrument and the serial number.
- Calibrated at 15 °C, A2LA Traceable hydrometers
- Graduated in kg/m³, accuracy: 0,5 kg/m³
- Reading below meniscus
Frequently Bought Together