Stem castors for shelving systems, Super Erecta®
About this item
Stem castors: Use with Super Erecta® posts and shelves to create a mobile shelving unit to meet special needs. Stem castors are shipped with donut bumper. 5mDA series provides extra shock absorption while the 5mP series is long-wearing and resists abrasion. 5m series is the most economical castor offered. 5mFA series are designed to dampen vibration and offer superior rollability and maneuverability.
ESD Classification: non-conductive.
Stainless steel cart-washable stem castors: Stem castors are shipped with donut bumpers.
Polymer castors: Innovative polymer stem castors offer corrosion resistance and enhanced durability. Appropriate for all medium-duty mobile applications. Donut bumpers included.
All-polymer horn, stainless steel axle and hardware
127 mm diameter polyurethane wheel, sleeve-style axle bearing and an optional toe-operated brake mechanism
Polyurethane, flat wheel tread.
29 Options Available Below
Product Details & Documents
Stem castors: Use with Super Erecta® posts and shelves to create a mobile shelving unit to meet special needs. Stem castors are shipped with donut bumper. 5mDA series provides extra shock absorption while the 5mP series is long-wearing and resists abrasion. 5m series is the most economical castor offered. 5mFA series are designed to dampen vibration and offer superior rollability and maneuverability.
ESD Classification: non-conductive.
Stainless steel cart-washable stem castors: Stem castors are shipped with donut bumpers.
Polymer castors: Innovative polymer stem castors offer corrosion resistance and enhanced durability. Appropriate for all medium-duty mobile applications. Donut bumpers included.
All-polymer horn, stainless steel axle and hardware
127 mm diameter polyurethane wheel, sleeve-style axle bearing and an optional toe-operated brake mechanism
Polyurethane, flat wheel tread.
Product Family Options
Stem casters
Product Information
- ForAvailabilityPrice
- METO4LDStem/swivel castor, resilient donut (Ø 102 mm)Stem/swivel castor, resilient donut (Ø 102 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 50.70
- METO5MBStem/brake castor, resilient, flat (Ø 127 mm)Stem/brake castor, resilient, flat (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 79.10
- METO5MBCStem/brake castor, resilient, flat (Ø 127 mm)Stem/brake castor, resilient, flat (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 127.00
- METO5MBESDStem/brake castor, conductive (Ø 127 mm)Stem/brake castor, conductive (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 154.00
- METO5MStem/swivel castor, resilient donut (Ø 127 mm)Stem/swivel castor, resilient donut (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 65.50
- METO5MCConductive stem/swivel castor, for ESD floor surface (Ø 127 mm)Conductive stem/swivel castor, for ESD floor surface (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 114.00
- METO5MDAStem/swivel castor, high modulus donut (Ø 127 mm)Stem/swivel castor, high modulus donut (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 76.00
- METO5MDBAStem/brake castor, high modulus donut (Ø 127 mm)Stem/brake castor, high modulus donut (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 87.70
- METO5MDRAStem/rigid castor, high modulus donut (Ø 127 mm)Stem/rigid castor, high modulus donut (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 87.70
- METO5MESDStem/swivel castor, conductive (Ø 127 mm)Stem/swivel castor, conductive (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 141.00
- METO5MFAStem/swivel castor, high modulus (Ø 127 mm)Stem/swivel castor, high modulus (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 117.00
- METO5MFBAStem/brake castor, high modulus (Ø 127 mm)Stem/brake castor, high modulus (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 131.00
- METO5MPBStem/brake castor, PA, flat (Ø 127 mm)Stem/brake castor, PA, flat (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 114.00
- METO5MPStem/swivel castor, PA, flat (Ø 127 mm)Stem/swivel castor, PA, flat (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 104.00
- METO6MPBStem/brake castor, PU, flat (Ø 152 mm)Stem/brake castor, PU, flat (Ø 152 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 160.00
- METO6MPRStem/rigid castor, PU, flat (Ø 152 mm)Stem/rigid castor, PU, flat (Ø 152 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 160.00
Stainless steel casters
Product Information
- ForAvailabilityPrice
- METO5MDBGSAStem/brake castor high modulus donut, (Ø 127 mm)Stem/brake castor high modulus donut, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 315.00
- METO5MDGSAStem/swivel castor high modulus donut, (Ø 127 mm)Stem/swivel castor high modulus donut, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 299.00
- METO5MPBGSAStem/brake castor, PA flat, (Ø 127 mm)Stem/brake castor, PA flat, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 328.00
- METO5MPGSAStem/swivel castor, PA flat, (Ø 127 mm)Stem/swivel castor, PA flat, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 317.00
- METO5MPRGSAStem/rigid castor, PU flat, (Ø 127 mm)Stem/rigid castor, PU flat, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 328.00
Polymer casters
Product Information
- ForAvailabilityPrice
- METO5PCBStem/brake polymer castor, (Ø 127 mm)Stem/brake polymer castor, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 170.00
- METO5PCBMStem/brake polymer and antimicrobial castor, (Ø 127 mm)Stem/brake polymer and antimicrobial castor, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 184.00
- METO5PCMStem/swivel polymer castor, (Ø 127 mm)Stem/swivel polymer castor, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 171.00
- METO5PCStem/swivel polymer and antimicrobial castor, (Ø 127 mm)Stem/swivel polymer and antimicrobial castor, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 154.00
High temperature autoclave stem casters
Product Information
- ForAvailabilityPrice
- METO5MHTNBStem/brake castor, PU flat, (Ø 127 mm)Stem/brake castor, PU flat, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 470.00
- METO5MHTNStem/swivel castor, PU flat, (Ø 127 mm)Stem/swivel castor, PU flat, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 444.00
- METO5MHTPBStem/brake castor, PU flat, (Ø 127 mm)Stem/brake castor, PU flat, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 470.00
- METO5MHTPStem/swivel castor, PU flat, (Ø 127 mm)Stem/swivel castor, PU flat, (Ø 127 mm)Super Erecta® shelvingLog in for availability1 * 1 STSFr. 444.00