Order Entry
Switzerland
ContactUsLinkComponent
Alkaline cleaners for machine washing of surgical instruments, Helimatic® Cleaner alcaline
Alkaline cleaners for machine washing of surgical instruments, Helimatic® Cleaner alcaline
Catalog # BRAU18731
Supplier:  B. Braun
undefined
Alkaline cleaners for machine washing of surgical instruments, Helimatic® Cleaner alcaline
Catalog # BRAU18731
Supplier:  B. Braun

Specifications

  • Capacity:
    5 l
  • Description:
    Helimatic® alkaline cleaner
  • Pk:
    1

Specifications

About this item

  • Effective cleaner of organic deposits
  • Typical dosage: 0,3 to 0,5%
  • Alkaline> pH10