Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Uridine 99%

Uridine 99%
Certificates
Uridine 99%

4 Options Available Below

Product Details & Documents
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
  • Melting Pt:162…170 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-407-5

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    162 to 170 °C
  • Loss on drying:
    ≤1 % (105 °C, 2 hrs)
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≥98.5 %
  • UV spectrophotometry:
    ≥98 % (lambda max. 262 nm, pH 7)
  • UV transmittance:
    ≥95 % (c=1, H2O, 430 nm) 1 cm cell
  • Ratio of Absorbances:
    0.72 to 0.76 (A250/A260)
  • Heavy metals (as Pb):
    ≤10 ppm
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 37.80
    SFr. 37.80
    1 * 1 g
       SFr. 37.80

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 231.00
    SFr. 231.00
    1 * 25 g
       SFr. 231.00

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 645.00
    SFr. 645.00
    1 * 100 g
       SFr. 645.00

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 2’120.00
    SFr. 2’120.00
    1 * 500 g
       SFr. 2’120.00