Order Entry
Switzerland
Orders LinkContactUsLinkComponent
DIPSO

DIPSO

Glass bottle for solids, 25 g
Catalog # ROTH7151.1
Marketsource
Supplier:   Roth Carl
DIPSO
Catalog # ROTH7151.1
Supplier:  Roth Carl
Marketsource
CAS Number:  68399-80-4
DIPSO

Glass bottle for solids, 25 g

CAS Number:  68399-80-4
Price
103.10 SFr.
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
  • CAS Number:68399-80-4
  • Formula:C₇H₁₇NO₆S
  • MDL Number:MFCD00038353
  • Molecular Weight:261.29 g/mol
  • Storage Temperature:Ambient
  • EINECS:269-992-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications