

4-Hydroxybenzenesulphonic acid 65% in aqueous solution, Sigma-Aldrich®
: Merck
:98-67-9
:
:C₆H₆O₄S
:MFCD00673723
:174.17 g/mol
:8,II
:M-Clarity™ quality level = MQ200
:202-691-6
:1803
:GHS05
:GHS08
:Ambient
- Solubility:(20 °C) soluble
- Boiling Point:100 °C
- Density:1.33 g/cm3 (20 °C)
- pH:1 (50 g/l, H2O, 20 °C)
- Synonyms:4-Hydroxybenzenesulfonic acid
Frequently Bought Together