ContactUsLinkComponent
Face screen without carrier, S591
Face screen without carrier, S591
Catalog # CENUS591
Face screen without carrier, S591
Catalog # CENUS591
Specifications
  • Type:
    Chemical resistant
  • Description:
    Face screen without carrier
  • Material:
    Acetate
  • Pk:
    1
Specifications
About this item
Long acetate face screen, to be combined with the carrier S54 for helmet attachment.
  • Resistant against organic and inorganic chemicals
  • Ideal for metal cutting applications
Length: 210 mm
Certifications: EN 166