Safety ankle boots, lace-up, Jaltarmac
Supplier: Jallatte
Certificates
About this item
Safety boots specifically for road construction. Made from hydrophobic nubuck leather with heat resistant Softane™/NBR (nitrile rubber) outer sole (to 300 °C).
- Confort-Tech integrated insole: Anatomical, antibacterial, shock absorber heel
- Isolating multi-layer complex insole prevents from heat shocks which may occur when working on asphalt or floors with temperatures of 160 °C
- Sole with lug tread for best anti-slip properties and improved shock absorption
- Protective steel toecap (200 J)
Colour: Black/red
Certifications: EN 345-1; S3, HI, HRO
Frequently Bought Together