Flame-retardant overalls with bib and braces, FLAM Bib'N'Brace
Trousers made of a cotton/polyester fabric with a special treatment, providing protection against heat and open flames. The trousers also feature fluorescent fabrics for an improved visibility.
- Two-way front zip
- Two loose-hanging, detachable chest pockets, one with flap and concealed snap fasteners, one with extra pocket and pen pocket
- Button and loop for sheath knife
- Two front pockets, two loose-hanging pockets with extra width, one with extra pocket, one with three tool pockets and tool loops, hammer loop
- Two back pockets with extra width, leg pocket with phone pocket and tool pocket
- Folding rule pocket over side seam, with tool pocket and button and loop for sheath knife
- Rrounded knees with knee pockets
Ideal for applications that require protection against flames, open arcs and welding sparks.
Weight: 365 g/m²
: EN 61482-1-2 (class 1); EN ISO 11612 (A1, B1, C1)
- Catalog No:
- KWIN100328540C60
- KWIN100328540C58
- KWIN100328540C62
- KWIN100328540C46
- KWIN100328540C50
- KWIN100328540C48
- KWIN100328540C52
- KWIN100328540C56
- KWIN100328540C54
- Model:
- FLAM Bib'N'Brace
- FLAM Bib'N'Brace
- FLAM Bib'N'Brace
- FLAM Bib'N'Brace
- FLAM Bib'N'Brace
- FLAM Bib'N'Brace
- FLAM Bib'N'Brace
- FLAM Bib'N'Brace
- FLAM Bib'N'Brace
- Garment application:
- Welding, Industrial
- Welding, Industrial
- Welding, Industrial
- Welding, Industrial
- Welding, Industrial
- Welding, Industrial
- Welding, Industrial
- Welding, Industrial
- Welding, Industrial
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Weight:
- 365 g/m²
- 365 g/m²
- 365 g/m²
- 365 g/m²
- 365 g/m²
- 365 g/m²
- 365 g/m²
- 365 g/m²
- 365 g/m²
Frequently Bought Together