Cut resistant gloves, Nitras® TAEKI5® 6720
Supplier: AS ARBEITSSCHUTZ
Certificates
About this item
Knitted glove made of TAEKI5® special cut protection fiber knit (13 mesh), with black latex coating on palm and fingertips.
- Latex coating for good grip and high acid resistance
- Good abrasion and high cut resistance, heat resistant
- Seamless processing
Ideal for cutting work in the metal and automotive industries where good touch is also required.
Farbe: grau
EN 388 4542
Certifications: EN 388 (3443C), EN 407 (X2XXXX); Cat. II
Specifications
- Catalog No:
- ASAS6720/S
- ASAS6720/M
- ASAS6720/XXL
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Gauge:
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (3443C), EN 407 (X2XXXX)
- EN 388 (3443C), EN 407 (X2XXXX)
- EN 388 (3443C), EN 407 (X2XXXX)