Electrical resistant gloves, Electrosoft CL 0
Supplier: HONEYWELL SAFETY
Certificates
About this item
Natural latex insulating gloves for electrical work with a maximum voltage of 1000 volts.
- Ergonomic shape and lightly powdered interior make the glove easy to slip on and off
- Natural latex base offers high dielectric characteristics
- Thickness ensures good dexterity while offering protection up to 1000 volts
Please note: Must be used with a leather overglove.
Thickness: 1 mm
Colour: Beige
EN 60903-2003, CEI 903-2002; Cat. III
Certifications: EN 60903-2003, CEI 903-2002; Cat. III
Specifications
- Catalog No:
- DALL2091907/11
- DALL2091907/8
- Model:
- CL 0
- CL 0
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Powdered / powder-free:
- Low-powdered
- Low-powdered
- Finish:
- Smooth
- Smooth
- Cuff style:
- Gauntlet
- Gauntlet
- EN norm:
- EN 60903-2003, CEI 903-2002
- EN 60903-2003, CEI 903-2002