Order Entry
Switzerland
ContactUsLinkComponent
Goggles, Muria
Goggles, Muria
Catalog # 111-0905
Supplier:  DELTA PLUS
Goggles, Muria
Catalog # 111-0905
Supplier:  DELTA PLUS

Specifications

  • Colour:
    Grey
  • Lenses:
    Clear PC
  • Pk:
    1

Specifications

About this item

These goggles have PVC transparent frames and PC lenses for protection against impacts caused by medium energy (impact 120 m/s), during grinding work.

  • Woven elastic band
  • Direct ventilation
  • Soft PVC nose bridge

Weight: 60 g

Certifications: Certified with EN 166 1 BT / BT.