Vials, Vacule®, WHEATON®
These vacule vials made of 33 low extractable clear borosilicate glass feature a heavy wall construction designed for lyophilisation and freeze-drying applications.
- Vials have controlled ID, for accurate stopper fit
- Screw threads eliminate use of crimp seal
: Meets USP Type I and ASTM E 438 Type I.
: Closures for 4 ml vials have to be ordred separately.
: Vials, 5 and 10 ml are supplied with unlined black phenolic cap.
Screw caps
Frequently Bought Together