Vibratory feeder DR 100 series
Supplier: Retsch GmbH
Certificates
About this item
The Vibratory feeder is used for uniform continuous feed of pourable bulk materials and fine powders.
- Reproducible results from uniform feed
- Digital time setting or continous operation
- Infinitely adjustable flow volume 0 to 5 litres/minute
- Easy to set up and operate
- Different sizes of feed shutes and hoppers to suit a a wide range of materials
Frequently Bought Together