TubingDialysis TubingDialysis tubing Spectra/Por® 5
MWCO 12 - 14 kD, regenerated cellulose
Supplied dry, preservative: glycerol
: PanReac AppliChem
: VWR Collection
: VWR Chemicals
: Sartorius Balances