Order Entry
Switzerland
ContactUsLinkComponent
Reagent digger, SP Bel-Art
Reagent digger, SP Bel-Art
  BELAF367130000
undefined
Reagent digger, SP Bel-Art
  BELAF367130000

 

  • Ø:
    24 mm
  • Length:
    229 mm
  • Material:
    Stainless steel
  • Pk:
    1

 

 

Stainless steel. This robust tool scrapes and scoops caked samples from all vessels.

  • Ideal for retrieving flaked, powdered or granular media
  • V-shape directs sample dispensing
  • Fits into bottles with minimum neck diameter of 24 mm