ContactUsLinkComponent
SP Bel-Art® tapered plug sampler
SP Bel-Art® tapered plug sampler
Catalog # BELAH378260000
undefined
SP Bel-Art® tapered plug sampler
Catalog # BELAH378260000
Specifications
  • Ø:
    17 mm
  • Description:
    Tapered plug sampler
  • Length:
    191 mm
  • Material:
    Stainless steel
  • Pk:
    1
Specifications
About this item
Tapered plug sampler can remove a sample from bags.
  • Sharp forward edge pierces packaging to retrieve sample
  • T-shape handle applies rotating force to cut sample
  • Easy cleaning and corrosion resistance
Suitable for blocks of cheese, grease, soap, clay, or any other similar semi-solids.