Specifications
- Catalog No:
- TEST0563.2051
- TEST0563.2052
- Model:
- 205
- 205
- pH range:
- 0,00...+14,00
- 0,00...+14,00
- pH resolution:
- 0,01
- 0,01
- pH accuracy:
- ±0,02
- ±0,02
- pH calibration:
- 1-, 2- or 3-point
- 1-, 2- or 3-point
- Automatic buffer recognition:
- Automatic full-scale value recognition
- Automatic full-scale value recognition
- Temperature range:
- 0...+60 °C (Short-term to +80 °C max. 5 min)
- 0...+60 °C (Short-term to +80 °C max. 5 min)
- Temperature resolution:
- 0,1 °C
- 0,1 °C
- Temperature accuracy:
- ±0,4 °C
- ±0,4 °C
- Display:
- Backlit, 2-line LCD
- Backlit, 2-line LCD
- Battery type:
- 4× Button cell LR44
- Battery life:
- 80 h (auto off 10 min)
- 80 h (auto off 10 min)
- Power supply:
- 4× Button cell LR44
- 4× Button cell LR44
- Operating temperature range:
- 0...+50 °C
- 0...+50 °C
- Storage temperature:
- −20...+70 °C
- −20...+70 °C
- IP rating:
- IP 65
- IP 65
- W×D×H:
- 145×38×167 mm
- 145×38×167 mm
- Weight:
- 0,135 kg
- 0,135 kg
Specifications
Related Information
CE Handheld pH Meter with Temperature Penetration Probe 205 CE pH Temperature Meter Handheld 205 testo-CE-Konformitaet_testo205ph1_0213-2017_D_GB testo-CE-Konformitaet_testo205ph1_0214-2017_D_GB
CoC Handheld pH Meter with Temperature Penetration Probe 205 CoC Products Coming into Contact with Food reg 1935 2004
Frequently Bought Together