Order Entry
Switzerland
ContactUsLinkComponent
 
Ribonuclease A
 :  EUROCLONE
 
Ribonuclease A
 :  EUROCLONE

 

  • CAS No.:
    9001-99-4
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00132184
  • Enzyme name:
    Ribonuclease A
  • Enzyme synonyms:
    RNase A

 

Frequently Bought Together