Order Entry
Switzerland
ContactUsLinkComponent
 
Desoxyribonuclease I
Catalog # EURLEMR126025
Supplier:  EUROCLONE
CAS Number:  
Desoxyribonuclease I
Catalog # EURLEMR126025
Supplier:  EUROCLONE
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • CAS No.:
    9003-98-9
  • Storage conditions:
    Freezer
  • Enzyme name:
    Desoxyribonuclease I
  • Enzyme synonyms:
    DNase I

Specifications

About this item