Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Electrophoresis Buffers
Running Buffers
Buffer, TRIS-tricine-SDS solution 10X concentrate
Buffer, TRIS-tricine-SDS solution 10X concentrate
Buffer, TRIS-tricine-SDS solution 10X concentrate
Catalog # NADIEC-869-1L
Supplier:
NATIONAL DIAGNOSTICS
CAS Number:
Expand image
Buffer, TRIS-tricine-SDS solution 10X concentrate
Catalog # NADIEC-869-1L
Supplier:
NATIONAL DIAGNOSTICS
CAS Number:
Specifications
Pack type:
Glass bottle
Pk:
1 L
Description:
TRIS-tricine-SDS buffer solution 10X concentrate
Specifications
About this item
Product Details
Product Details