Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Buffer, TE solution (TRIS-EDTA buffer) pH 7,5

TE buffer solution (TRIS-EDTA buffer) pH 7,5, Glass bottle, 1 L
Catalog # MOLE55376245-1L
Marketsource
Supplier:   Molekula
Buffer, TE solution (TRIS-EDTA buffer) pH 7,5
Catalog # MOLE55376245-1L
Supplier:  Molekula
Marketsource
CAS Number:  
Buffer, TE solution (TRIS-EDTA buffer) pH 7,5

TE buffer solution (TRIS-EDTA buffer) pH 7,5, Glass bottle, 1 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TE buffer solution (TRIS-EDTA buffer) pH 7,5

Specifications