Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer)

Buffer, TE solution 100x concentrate (TRIS-EDTA buffer)

TE buffer solution 100x concentrate (TRIS-EDTA buffer), Glass bottle, 25 mL
Catalog # NADIEC-862-25ML
Marketsource
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer)
Catalog # NADIEC-862-25ML
Marketsource
CAS Number:  
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer)

TE buffer solution 100x concentrate (TRIS-EDTA buffer), Glass bottle, 25 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 25 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Description:
    TE buffer solution 100x concentrate (TRIS-EDTA buffer)

Specifications