Order Entry
Switzerland
ContactUsLinkComponent
Hygromycin B
Toxic
Corrosive
Health
Hygromycin B
Catalog # MOLE38888840-100MG
Supplier:  Molekula
CAS Number:  
Toxic
Corrosive
Health
Hygromycin B
Catalog # MOLE38888840-100MG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :527.5 g/mol
Storage Temperature:Refrigerator
EINECS:250-545-5
UN:3462
ADR:6.1,I
 :GHS06
 :GHS05
 :GHS08
CAS Number:31282-04-9
Formula:C₂₀H₃₇N₃O₁₃
MDL Number:MFCD06795479