Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hygromycin B
Hygromycin B
Hygromycin B
A5347.0100
:
PanReac AppliChem
:
Hygromycin B
A5347.0100
:
PanReac AppliChem
:
Pack type:
Glass bottle for solids
Pk:
100 mg
Seq:
2440
:
:
527.5 g/mol
:
Refrigerator
:
250-545-5
:
3462
:
6.1,I
:
GHS06
:
GHS05
:
GHS08
:
31282-04-9
:
C₂₀H₃₇N₃O₁₃
:
MFCD06795479