Order Entry
Switzerland
ContactUsLinkComponent
Dispensing cannula for 5, 10 and 20 ml Dosys™ syringes, SOCOREX® DOSYS™
Dispensing cannula for 5, 10 and 20 ml Dosys™ syringes, SOCOREX® DOSYS™
Catalog # WHEA851412
Supplier:  DWK Life Sciences
undefined
Dispensing cannula for 5, 10 and 20 ml Dosys™ syringes, SOCOREX® DOSYS™
Catalog # WHEA851412
Supplier:  DWK Life Sciences

Specifications

  • Ø×L:
    2,2×100 mm
  • Description:
    Dispensing cannula with Luer Lock, for 5, 10 and 20 mLlDosys™ syringes
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

Blunt end dispensing cannulas with Luer Lock. Ideal for use with 24 and 96 well microplates.

  • Stainless steel
  • Autoclavable

This unique dispensing system enables you to perform sample inoculations and cell feedings faster and more efficiently under sterile conditions.