Order Entry
Switzerland
ContactUsLinkComponent
Contact plate racks, SP Bel-Art
Contact plate racks, SP Bel-Art
Codice catalogo BELAF189900000
Nome del fornitore:  Bel-Art Products, a Part of SP
undefined
Contact plate racks, SP Bel-Art
Codice catalogo BELAF189900000
Nome del fornitore:  Bel-Art Products, a Part of SP

Specifiche

  • Description:
    Contact plate rack, 266×165×178 mm
  • Height:
    178 mm
  • Pk:
    1

Specifiche

Informazioni su questo articolo

PP moulded rack, ideal for high volume sampling processes.

  • Transport and incubate contact plates and organise bench top sample processing
  • Racks hold 60 contact plates with covers, configured in six columns of ten plates
  • Interlocking when stacked
  • Large centre divider serves as a handle that does not interfere with samples and provides space for labelling
  • Autoclavable at 121 °C

Some simple assembly required. Dimensions provide stability and conserve bench space: 266×165×178 mm.

Four fully loaded racks can be safely stacked to maximise incubator space.

Access slots underneath each stack permits secure removal of a complete stack of plates from the rack using a fixed grip that places a finger under the bottom plate and the thumb on top of the cover of the top plate.