Contact plate racks, Poxygrid®, SP Bel-Art
Epoxy coated steel wire. Holds 10 or 30 contact plates with lids arranged in one or three stacks of ten plates.
- Prevents accidents during transport
- Chemical and corrosion resistant
- Compact size maximises incubator space and won't crowd bench top
W×D×H: 82×82×140 cm (one stack)
W×D×H: 246×82×140 mm (three stacks)
Frequently Bought Together