ContactUsLinkComponent
Pestle with extended handle, Bel-Art ProCulture, SP Bel-Art
Pestle with extended handle, Bel-Art ProCulture, SP Bel-Art
Catalog # 212-7310
undefined
Pestle with extended handle, Bel-Art ProCulture, SP Bel-Art
Catalog # 212-7310
Specifications
  • Description:
    For 1,5 ml microcentrifuge tubes, with rounded tip cone
  • Length:
    210 mm
  • Environmentally Preferable:
  • Pk:
    6
Specifications
About this item
Polypropylene tissue grinder efficiently homogenizes tissue and cells in microcentrifuge, centrifuge, and grinding tubes.
  • Polypropylene construction
  • Rounded end design
  • Tapered conical tip
The cone tip tapers from 8,2 mm (0,33") down to 2,9 mm (0,12") in a length of 19,8 mm (0,78"). It features a rounded end. Overall length is 21 cm (8¼"), with a 4,7 mm (³/₁₆") diameter shaft. Steam autoclavable at 121 °C (250 °F).