ContactUsLinkComponent
VWR®, Destaining bags
VWR®, Destaining bags
Catalog # E732-25
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Destaining bags
Catalog # E732-25
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    25
  • Description:
    Destaining bags
Specifications
About this item
Destaining bags are designed to safely remove ethidium bromide from solution during overnight treatment. Each bag extracts up to 5 mg of ethidium bromide from solution, thus minimising the exposure of research personnel to this toxic material. The special absorbent mixture retains dye molecules in a convenient bag for removal and incineration.
  • Removes up to 5 mg ethidium bromide per bag
  • Minimises exposure to toxic materials
  • Easiest ethidium bromide removal available