Hollow fiber filter modules, MicroKros®
Supplier: Spectrum Laboratories
Certificates
About this item
Designed for gentle tangential flow filtration of small volumes ranging from 1 to 100 ml. MicroKros® modules are typically operated by a peristaltic pump, these devices also offer the unique advantage of manual operation for quick and convenient separations for small volumes.
- 5 to 30 minute separations
- Two modes: Pump operation and Manual operation
- Gentle process for increased viability
- 90 to 99% product purity and 85 to 95% product recovery
Delivery: The syringe accessory kit includes 10 × 5 ml syringes, 15 × male Luer cap, 10 × male Luer to 3/16" hose barb fitting, 5 × check valve, 10 × female Luer T connector.
Specifications
- Catalog No:
- SPECC02-E010-10-S
- SPECC04-S010-05-P
- SPECC02-M10U-06-N
- SPECC02-E030-05-N
- SPECC04-S500-05-S
- SPECC04-S050-05-N
- SPECC02-P20U-10-N
- SPECC02-P20U-05-N
- SPECC02-P20U-05-S
- SPECC02-E500-10-S
- SPECC02-E050-10-S
- SPECC02-E500-05-S
- SPECC02-M20U-06-S
- SPECC02-M20U-10-N
- SPECC02-M20U-10-S
- SPECC02-E500-10-N
- SPECC04-S010-05-S
- SPECC06-S010-05-N
- SPECC06-S010-05-S
- SPECC06-S050-05-N
- SPECC06-S010-05-P
- SPECC02-E500-05-N
- SPECC02-S500-05-S
- SPECC02-E010-05-N
- SPECC02-E100-10-N
- SPECC02-M20U-06-N
- SPECC02-S050-05-P
- SPECC02-M10U-06-S
- SPECC02-E300-05-S
- SPECC02-E300-10-N
- SPECC02-E300-10-S
- SPECC02-E010-10-N
- SPECC02-E100-05-N
- SPECC02-E030-05-S
- SPECC06-S500-05-N
- SPECC02-S05U-05-P
- SPECC02-S05U-05-S
- SPECC02-S500-05-N
- SPECC06-S050-05-P
- SPECC02-S500-05-P
- SPECC02-S050-05-N
- SPECC02-S050-05-S
- SPECC04-S05U-05-P
- SPECC04-S500-05-N
- SPECC04-S010-05-N
- SPECC02-S010-05-P
- SPECC02-S010-05-S
- SPECC02-E010-05-S
- SPECC02-E050-05-S
- SPECC02-E100-05-S
- SPECC02-S010-05-N
- SPECC02-S05U-05-N
- SPECC04-S500-05-P
- SPECC04-S050-05-S
- SPECC02-P20U-10-S
- SPECC02-E005-05-S
- SPECC02-E070-10-N
- SPECC02-E070-10-S
- SPECC02-E050-10-N
- SPECC02-E003-05-N
- SPECC02-E100-10-S
- SPECC02-E070-05-N
- SPECC02-E003-05-S
- SPECC02-E005-05-N
- SPECC02-E070-05-S
- SPECC02-E030-10-N
- SPECC06-S500-05-S
- SPECC06-S050-05-S
- SPECC02-E030-10-S
- SPECC02-E050-05-N
- SPECC02-E300-05-N
- SPECC06-S05U-05-P
- SPECC06-S500-05-P
- SPECC04-S050-05-P
- Ein-/Auslass Verbindungen:
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock
- Female Luer lock/male Luer lock