Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Abietic acid

Abietic acid

Glass bottle for solids, 1 kg
Catalog # ROTH5005.1
Marketsource
Supplier:   Roth Carl
Abietic acid
Catalog # ROTH5005.1
Supplier:  Roth Carl
Marketsource
CAS Number:  514-10-3
Abietic acid

Glass bottle for solids, 1 kg

CAS Number:  514-10-3
Price
SFr. 65.90
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:514-10-3
  • Formula:C₂₀H₃₀O₂
  • MDL Number:MFCD03423567
  • Molecular Weight:302.45 g/mol
  • EINECS:208-178-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.