Abietic acid ≥75%, tech.
Supplier: Thermo Fisher Scientific
About this item
Purity: 75% CAS No.: 514-10-3 Molecular Formula: C20H30O2 Formula Weight: 302.45 Storage Temperature: 2°C to 8°C Odour: Characteristic Beilstein Reference: 2221451 Merck Reference: 14,7 MDL No.: MFCD03423567
- CAS Number:514-10-3
- Formula:C₂₀H₃₀O₂
- MDL Number:MFCD03423567
- Molecular Weight:302.45 g/mol
- Merck Index:14,7
- Storage Temperature:Refrigerator
- EINECS:208-178-3
2 Options Available Below
Product Details & Documents
Purity: 75% CAS No.: 514-10-3 Molecular Formula: C20H30O2 Formula Weight: 302.45 Storage Temperature: 2°C to 8°C Odour: Characteristic Beilstein Reference: 2221451 Merck Reference: 14,7 MDL No.: MFCD03423567
- CAS Number:514-10-3
- Formula:C₂₀H₃₀O₂
- MDL Number:MFCD03423567
- Molecular Weight:302.45 g/mol
- Merck Index:14,7
- Storage Temperature:Refrigerator
- EINECS:208-178-3



