Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Hydroxyectoine

Hydroxyectoine

Glass bottle for solids, 100 g
Catalog # BMOL55460-4
Supplier:   Biomol Enzo
Hydroxyectoine
Catalog # BMOL55460-4
Supplier:  Biomol Enzo
CAS Number:  165542-15-4
Hydroxyectoine

Glass bottle for solids, 100 g

CAS Number:  165542-15-4
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:165542-15-4
  • Formula:C₆H₁₀N₂O₃
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.