Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Ectoine

Glass bottle for solids, 1 g
Catalog # BMOL53238-1
Supplier:   Biomol Enzo
Ectoine
Catalog # BMOL53238-1
Supplier:  Biomol Enzo
CAS Number:  96702-03-3
Ectoine

Glass bottle for solids, 1 g

CAS Number:  96702-03-3
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic acid
  • CAS Number:96702-03-3
  • Formula:C₆H₁₀N₂O₂
  • MDL Number:MFCD03419286
  • Molecular Weight:142.1558 g/mol
  • EINECS:431-910-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.