Pesticide reference standard, Fenvalerate 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL13630000IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:51630-58-1
- Description:Fenvalerate 10 µg/ml in isooctane
- Formula:C₂₅H₂₂ClNO₃
- Description:Fenvalerat