Order Entry
Switzerland
Orders LinkContactUsLinkComponent
(2-Hydroxylpropyl)-β-cyclodextrine

(2-Hydroxylpropyl)-β-cyclodextrine

Glass bottle for solids, 25 g
Catalog # MOLE51779232-25G
Marketsource
Supplier:   Molekula
(2-Hydroxylpropyl)-β-cyclodextrine
Catalog # MOLE51779232-25G
Supplier:  Molekula
Marketsource
CAS Number:  128446-35-5
(2-Hydroxylpropyl)-β-cyclodextrine

Glass bottle for solids, 25 g

CAS Number:  128446-35-5
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
  • CAS Number:128446-35-5
  • Formula:C₆₃H₁₁₂O₄₂
  • MDL Number:MFCD16621721
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications