4-Bromophenyl phenyl ether (BDE No. 3)
Catalog # EHERC15898003
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
- Environmentally Preferable:
Specifications
About this item
Synonyms: 4-Bromodiphenyl ether, 1-Bromo-4-phenoxybenzene
:
:249.11 g/mol
EINECS:202-952-4
UN:3082
CAS Number:101-55-3
Formula:C₁₂H₉BrO
MDL Number:MFCD00000094
ADR:9 III