3,6-Dioxaoctamethylenediamine for synthesis, Sigma-Aldrich®
: Merck
:929-59-9
:
:C₆H₁₆N₂O₂
:MFCD00040474
:148.20 g/mol
:8,II
:M-Clarity™ quality level = MQ200
:213-203-6
:2735
:GHS05
:GHS07
:Ambient
- Solubility:(20 °C) soluble
- Molar Mass:148.20 g/mol
- Boiling Point:88 °C (0.7 hPa)
- Flash Point:121 °C
- Density:1.01 g/cm3 (20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Frequently Bought Together