Order Entry
Switzerland
ContactUsLinkComponent
Monolinuron (N-(4-Chlorophenyl)-N'-methoxy-N'-methylurea), Supelco®
Monolinuron (N-(4-Chlorophenyl)-N'-methoxy-N'-methylurea), Supelco®
Catalog # 45590-250MG
Supplier:  Merck
CAS Number:  
undefined
Monolinuron (N-(4-Chlorophenyl)-N'-methoxy-N'-methylurea), Supelco®
Catalog # 45590-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
EINECS:217-129-5
CAS Number:1746-81-2
Formula:C₉H₁₁ClN₂O₂
MDL Number:MFCD00055264