Monolinuron (N-(4-Chlorophenyl)-N'-methoxy-N'-methylurea)
Catalog # EHERC15310000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
EINECS:217-129-5
CAS Number:1746-81-2
Formula:C₉H₁₁ClN₂O₂
MDL Number:MFCD00055264